Anti-CSN1S1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BS-8245R-A488
BOSSBS-8245R-A488EA 890 EUR
BOSSBS-8245R-A488
Anti-CSN1S1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Important role in the capacity of milk to transport calcium phosphate.Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSN1S1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR