Anti-GTF2H1 Rabbit Polyclonal Antibody (Cy3®)

Toimittaja: Bioss
BS-8175R-CY3
BOSSBS-8175R-CY3EA 900 EUR
BOSSBS-8175R-CY3
Anti-GTF2H1 Rabbit Polyclonal Antibody (Cy3®)
Vasta-aineet
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTF2H1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR