Anti-GGPS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: Bioss
BS-8039R-HRP
BOSSBS-8039R-HRPEA 900 EUR
BOSSBS-8039R-HRP
Anti-GGPS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, both protein-coding and non-protein-coding, have been found for this gene. [provided by RefSeq, Sep 2010].
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500