Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Toimittaja: Bioss
BS-7949R-A555
BOSSBS-7949R-A555EA 893 EUR
BOSSBS-7949R-A555
Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Vasta-aineet
Catalyzes the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumor formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR