Anti-CCNDBP1 Rabbit Polyclonal Antibody (Cy5.5®)

Toimittaja: Bioss
BS-7885R-CY5.5
BOSSBS-7885R-CY5.5EA 900 EUR
BOSSBS-7885R-CY5.5
Anti-CCNDBP1 Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
May negatively regulate cell cycle progression. May act at least in part via inhibition of the cyclin-D1/CDK4 complex, thereby preventing phosphorylation of RB1 and blocking E2F-dependent transcription.Tissue specificity: Ubiquitously expressed. Expression is down-regulated in a variety of tumor types including breast, colon, prostate and rectal tumors, and is up-regulated in certain hepatic carcinomas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNDBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR