Anti-DLL3 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BS-7860R
BOSSBS-7860REA 725 EUR
BOSSBS-7860R
Anti-DLL3 Rabbit Polyclonal Antibody
Vasta-aineet
Delta-like 3 (DLL3) is a transmembrane Delta-like protein that inhibits primary neurogenesis. It may be required to divert neurons along a specific differentiation pathway and plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 is one of five DSL proteins that bind to the Notch receptor and activates Notch signaling.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DLL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR