Anti-KATNB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Toimittaja: Bioss
BS-7851R-FITC
BOSSBS-7851R-FITCEA 900 EUR
BOSSBS-7851R-FITC
Anti-KATNB1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Microtubules, polymers of alpha and beta tubulin subunits,form the mitotic spindle of a dividing cell and help to organizemembranous organelles during interphase. Katanin is a heterodimerthat consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDaaccessory protein (p80 subunit B 1). The p60 subunit acts to severand disassemble microtubules, while the p80 subunit targets theenzyme to the centrosome. Katanin is a member of the AAA family ofATPases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KATNB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (FluoresceinMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR