Anti-KLHL9 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BS-7753R
BOSSBS-7753REA 725 EUR
BOSSBS-7753R
Anti-KLHL9 Rabbit Polyclonal Antibody
Vasta-aineet
KLHL9 is the substrate-specific adapter for a CUL3-based E3 ubiquitin-protein ligase complex. Within this complex, KLHL9 controls the dynamic behavior of AURKB on mitotic chromosomes and thereby coordinates faithful mitotic progression and completion of cytokinesis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: KLHL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR