Anti-PDGFA / PDGFB Rabbit Polyclonal Antibody (Cy5.5®)

Toimittaja: Bioss
BS-7559R-CY5.5
BOSSBS-7559R-CY5.5EA 893 EUR
BOSSBS-7559R-CY5.5
Anti-PDGFA / PDGFB Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
Platelet-Derived Growth Factor (PDGF) is the major serum mitogen for cells of mesenchymal origin in humans. The biologically active protein is a dimer composed of two related polypeptides designated A and B. The PDGF protein has been implicated both directly as well as indirectly in several pathological states including neoplasia, arthritis, arteriosclerosis and bone marrow sclerosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDGFA / PDGFB
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR