Anti-SPON1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-7520R-A750
BOSSBS-7520R-A750EA 893 EUR
BOSSBS-7520R-A750
Anti-SPON1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Cell adhesion protein that promotes the attachment of spinal cord and sensory neuron cells and the outgrowth of neurites in vitro. May contribute to the growth and guidance of axons in both the spinal cord and the PNS (By similarity). Major factor for vascular smooth muscle cell.

Type: Primary
Antigen: SPON1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 741-807/807
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR