Anti-OAS3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Toimittaja: Bioss
BS-7344R-A488
BOSSBS-7344R-A488EA 893 EUR
BOSSBS-7344R-A488
Anti-OAS3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
The 2'- 5'- oligoadenylate synthetase (OAS) family is comprised of four members: OAS1, OAS2, OAS3 and OASL. These proteins are induced by interferons and function to convert ATP into 2'- 5'- linked oligomers of adenosine in the presence of double-stranded RNA and magnesium ions. Copper, iron and zinc ions strongly inhibit the OAS enzymatic activity, while manganese ions can replace magnesium ions as an activator. The OAS family plays a significant role in the inhibition of cellular protein synthesis, apoptosis and growth, and its members are important factors in viral infection resistance.More Product Information