Anti-SH3GLB1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Toimittaja: Bioss
BS-7097R-A488
BOSSBS-7097R-A488EA 893 EUR
BOSSBS-7097R-A488
Anti-SH3GLB1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
May be required for normal outer mitochondrial membrane dynamics. Required for coatomer-mediated retrograde transport in certain cells. May recruit other proteins to membranes with high curvature. May promote membrane fusion.Tissue specificity: Highly expressed in heart, skeletal muscle, kidney and placenta. Detected at lower levels in brain, colon, thymus, spleen, liver, small intestine, lung and peripheral blood leukocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SH3GLB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR