Anti-SDCCAG8 Rabbit Polyclonal Antibody

Toimittaja: Bioss
BS-7011R
BOSSBS-7011REA 725 EUR
BOSSBS-7011R
Anti-SDCCAG8 Rabbit Polyclonal Antibody
Vasta-aineet
The exact function of SDCCAG8 remains unknown. It is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa, colon and renal cancer tumors.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR