Anti-MYF5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-6936R-A750
BOSSBS-6936R-A750EA 893 EUR
BOSSBS-6936R-A750
Anti-MYF5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Differentiation of myogenic cells is regulated by multiple positively and negatively acting factors. One well characterised family of helix-loop-helix (HLH) proteins known to play an important role in the regulation of muscle cell development include Myo D, myogenin, Myf-5 and Myf-6 (also designated MRF-4 or herculin). Of interest, most muscle cells express either Myo D or Myf-5 in the committed state, but when induced to differentiate, all turn on expression of myogenin. Myo D transcription factors form heterodimers with products of a more widely expressed family of bHLH genes, the E family,More Product Information