Anti-MAGEA5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Toimittaja: Bioss
BS-6820R-FITC
BOSSBS-6820R-FITCEA 900 EUR
BOSSBS-6820R-FITC
Anti-MAGEA5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
MAGEA5 is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. This MAGEA gene encodes a protein that is C-terminally truncated compared to other family members,More Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli MAGEA5
Environmentally Preferable
Antigeenin nimi Melanoma Antigen Family A5
Antigeenin synonyymit MAGE5|MAGEA4|CT1.5
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio FITC (Fluorescein Isothiocyanate)
Reaktiivisuus Human
Isäntä Rabbit
Geeni ID 4104
Isotyyppi IgG
ImmunoKemia Yes
Lomake liquid
Puhdistus Purified by Protein A
Pitoisuus 1 μg/μl
Ristiadsorptio No
Säilytyspuskuri Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Säilytyslämpötila Store at 4°C for 12 months
Kuljetuslämpötila 4°C