Anti-Sertad1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-6691R-A750
BOSSBS-6691R-A750EA 893 EUR
BOSSBS-6691R-A750
Anti-Sertad1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Acts at E2F-responsive promoters as coregulator to integrate signals provided by PHD- and/or bromodomain-containing transcription factors. Stimulates E2F1/TFDP1 transcriptional activity. Renders the activity of cyclin D1/CDK4 resistant to the inhibitory effects of CDKN2A/p16INK4A.

Type: Primary
Antigen: Sertad1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-42/236
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

TEKNISET TIEDOT

Antigeenin nimi Sertad1
Environmentally Preferable
Antigeenin synonyymit SERTAD1|SERTA domain-containing protein 1|SEI1|Transcriptional regulator interacting with the PHD-bromodomain 1|CDK4-binding protein p34SEI1|TRIP-Br1|SEI-1
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Alexa Fluor® 750
Reaktiivisuus Mouse
Isäntä Rabbit
Geeni ID 29950
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Modification Unmodified
Epitooppi 1-42/236
Immunogeeni KLH conjugated synthetic peptide derived from human Sertad1
Lomake Liquid
Molekyylipaino 25 kDa
Puhdistus Purified by Protein A
Pitoisuus 1 µg/µl
Säilytyspuskuri Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Säilytyslämpötila Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Kuljetuslämpötila 4 °C
Testatut aplikaatiot ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR