Anti-STMN3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Toimittaja: Bioss
BS-6596R-A555
BOSSBS-6596R-A555EA 893 EUR
BOSSBS-6596R-A555
Anti-STMN3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Vasta-aineet
STMN3 belongs to the stathmin/oncoprotein 18 family of microtubule-destabilizing phosphoproteins. It is similar to the SCG10 protein and is involved in signal transduction and regulation of microtubule dynamics.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STMN3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR