Anti-KCNJ8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Toimittaja: Bioss
BS-6468R-A647
BOSSBS-6468R-A647EA 893 EUR
BOSSBS-6468R-A647
Anti-KCNJ8 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by external barium (By similarity).
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1More Product Information