Anti-C3 Rabbit Polyclonal Antibody (Cy5®)
Toimittaja: Bioss
BS-6416R-CY5
BOSSBS-6416R-CY5EA 922 EUR
BOSSBS-6416R-CY5
Anti-C3 Rabbit Polyclonal Antibody (Cy5®)
Vasta-aineet
C3 plays a central role in the activation of the complement system. Its processing by C3 convertase is the central reaction in both classical and alternative complement pathways. After activation C3b can bind covalently, via its reactive thioester, to cell surface carbohydrates or immune aggregates. Derived from proteolytic degradation of complement C3, C3a anaphylatoxin is a mediator of local inflammatory process. In chronic inflammation, acts as a chemoattractant for neutrophils (By similarity). It induces the contraction of smooth muscle, increases vascular permeability and causesMore Product Information