Anti-CDK5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Toimittaja: Bioss
BS-6411R-A350
BOSSBS-6411R-A350EA 895 EUR
BOSSBS-6411R-A350
Anti-CDK5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Vasta-aineet
CDK5 (Cyclin Dependent Kinase 5) is serine/threonine kinase involved in synaptic regulation and neuronal development; phosphorylates synaptic protein Pctaire1; regulates acetylcholine receptor expression. CDK5 is a member of the cyclindependent kinase family of serine/threonine kinases. It is present in numerous mammalian tissues including kidney, testes, and ovary. Its activity is detected almost exclusively in brain extracts. Neuronal and muscle cells contain the highest amount of this protein. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity, but Cdk5 is not activatedMore Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli CDK5
Environmentally Preferable
Antigeenin nimi Cyclin-dependent Kinase 5
Antigeenin synonyymit PSSALRE|TPKII catalytic subunit|TAU protein kinase II catalytic subunit|Cell division protein kinase 5|CRK6|CDK5|EC 2.7.11.22|kinase Cdk5|Tau protein kinase II catalytic subunit
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Alexa Fluor® 350
Reaktiivisuus Human,Rat,Mouse
Isäntä Rabbit
Geeni ID 1020
Isotyyppi IgG
ImmunoKemia Yes
Modification phospho-Tyr15
Lomake liquid
Puhdistus Purified by Protein A
Pitoisuus 1 μg/μl
Ristiadsorptio No
Säilytyspuskuri Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Säilytyslämpötila Store at 4°C for 12 months
Kuljetuslämpötila 4°C