Anti-pan methyl Lysine Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Toimittaja: Bioss
BS-6350R-A647
BOSSBS-6350R-A647EA 893 EUR
BOSSBS-6350R-A647
Anti-pan methyl Lysine Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
Lysine methylation occurs in three distinct states, having either one (me1), two (me2) or three (me3) methyl groups attached to the amine group of the lysine side chain. In eukaryotes, histone H3 trimethylated at lysine 4 (H3K4me3) is associated with active chromatin and gene expression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: pan methyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR