Anti-TCF3 Thr355 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Toimittaja: Bioss
BS-6066R-A680
BOSSBS-6066R-A680EA 1350 EUR
BOSSBS-6066R-A680
Anti-TCF3 Thr355 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Heterodimers between TCF3 and tissue-specific basic helix-loop-helix (bHLH) proteins play major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Dimers bind DNA on E-box motifs: 5'-CANNTG-3'. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer.

Type: Primary
Antigen: TCF3 Thr355
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/651
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR