Anti-RAD54B Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Toimittaja: Bioss
BS-5940R-A680
BOSSBS-5940R-A680EA 890 EUR
BOSSBS-5940R-A680
Anti-RAD54B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
The protein encoded by this gene belongs to the DEAD-like helicase superfamily. It shares similarity with Saccharomyces cerevisiae RAD54 and RDH54, both of which are involved in homologous recombination and repair of DNA. This protein binds to double-stranded DNA, and displays ATPase activity in the presence of DNA. This gene is highly expressed in testis and spleen, which suggests active roles in meiotic and mitotic recombination. Homozygous mutations of this gene were observed in primary lymphoma and colon cancer.

Type: Primary
Antigen: RAD54B
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR