Anti-OS9 Rabbit Polyclonal Antibody (Cy5®)

Toimittaja: Bioss
BS-5901R-CY5
BOSSBS-5901R-CY5EA 900 EUR
BOSSBS-5901R-CY5
Anti-OS9 Rabbit Polyclonal Antibody (Cy5®)
Vasta-aineet
Lectin which functions in endoplasmic reticulum (ER) quality control and ER-associated degradation (ERAD). May bind terminally misfolded non-glycosylated proteins as well as improperly folded glycoproteins, retain them in the ER, and possibly transfer them to the ubiquitination machinery and promote their degradation. Possible targets include TRPV4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OS9
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR