Anti-BCL2L12 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-5797R-A750
BOSSBS-5797R-A750EA 890 EUR
BOSSBS-5797R-A750
Anti-BCL2L12 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Expressed mainly in breast, thymus, prostate, foetal liver, colon, placenta, pancreas, small intestine, spinal cord, kidney, and bone marrow and to a lesser extent in many other tissues. Isoform 2 is primarily expressed in skeletal muscle.

Type: Primary
Antigen: BCL2L12
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/334
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR