Anti-SUV39H1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-5708R-A750
BOSSBS-5708R-A750EA 890 EUR
BOSSBS-5708R-A750
Anti-SUV39H1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
SUV39H1 is targeted to histone H3 via its interaction with RB1 and is involved in many processes, such as repression of MYOD1-stimulated differentiation, regulation of the control switch for exiting the cell cycle and entering differentiation, repression by the PML-RARA fusion protein, BMP-induced repression, repression of switch recombination to IgA and regulation of telomere length. Component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. TheMore Product Information