Anti-IDI1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Toimittaja: Bioss
BS-5073R-A647
BOSSBS-5073R-A647EA 893 EUR
BOSSBS-5073R-A647
Anti-IDI1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
IDI1 is a peroxisomally localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP). These are substrates for the reactions that ultimately result in the synthesis of cholesterol. There is reduction in IPP isomerase activity in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDI1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR