Anti-HIV1 p55+p17 Rabbit Polyclonal Antibody (Cy5.5®)
Toimittaja: Bioss
BS-4982R-CY5.5
BOSSBS-4982R-CY5.5EA 900 EUR
BOSSBS-4982R-CY5.5
Anti-HIV1 p55+p17 Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
HIV1 performs highly complex orchestrated tasks during the assembly, budding, maturation and infection stages of the viral replication cycle. During viral assembly, the proteins form membrane associations and self-associations that ultimately result in budding of an immature virion from the infected cell. Gag precursors also function during viral assembly to selectively bind and package two plus strands of genomic RNA. HIV1 Gag p17, the HIV1 matrix protein is produced by the digestion of its precursor Gag p55 by HIV1 protease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary