Anti-HIV1 p55+p17 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Toimittaja: Bioss
BS-4982R-A647
BOSSBS-4982R-A647EA 893 EUR
BOSSBS-4982R-A647
Anti-HIV1 p55+p17 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Vasta-aineet
HIV1 performs highly complex orchestrated tasks during the assembly, budding, maturation and infection stages of the viral replication cycle. During viral assembly, the proteins form membrane associations and self-associations that ultimately result in budding of an immature virion from the infected cell. Gag precursors also function during viral assembly to selectively bind and package two plus strands of genomic RNA. HIV1 Gag p17, the HIV1 matrix protein is produced by the digestion of its precursor Gag p55 by HIV1 protease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary