Anti-NS4B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Toimittaja: Bioss
BS-4857R-A488
BOSSBS-4857R-A488EA 893 EUR
BOSSBS-4857R-A488
Anti-NS4B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary