Anti-AQP2 Rabbit Polyclonal Antibody (Cy5.5®)

Toimittaja: Bioss
BS-4611R-CY5.5
BOSSBS-4611R-CY5.5EA 900 EUR
BOSSBS-4611R-CY5.5
Anti-AQP2 Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
Forms a water-specific channel that provides the plasma membranes of renal collecting duct with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR