Anti-EGF Rabbit Polyclonal Antibody (Cy5®)

Toimittaja: Bioss
BS-4568R-CY5
BOSSBS-4568R-CY5EA 909 EUR
BOSSBS-4568R-CY5
Anti-EGF Rabbit Polyclonal Antibody (Cy5®)
Vasta-aineet
EGF stimulates the growth of various epidermal and epithelial tissues in vivo and in vitro and of some fibroblasts in cell culture. Magnesiotropic hormone that stimulates magnesium reabsorption in the renal distal convoluted tubule via engagement of EGFR and activation of the magnesium channel TRPM6 (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR