Anti-Selenium Binding Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-4200R-A680
BOSSBS-4200R-A680EA 890 EUR
BOSSBS-4200R-A680
Anti-Selenium Binding Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colourectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.
Type: Primary
Antigen: SeleniumMore Product Information