Anti-KHK Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Toimittaja: Bioss
BS-4045R-A350
BOSSBS-4045R-A350EA 893 EUR
BOSSBS-4045R-A350
Anti-KHK Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Vasta-aineet
KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose 1 phosphate. The splice variant presented encodes the highly active form found in liver, renal cortex, and small intestine, while the alternate variant encodes the lower activity form found in most other tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KHK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR