Anti-TPI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Bioss
BS-4042R-HRP
BOSSBS-4042R-HRPEA 909 EUR
BOSSBS-4042R-HRP
Anti-TPI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
This gene encodes an enzyme, consisting of two identical proteins, which catalyzes the isomerization of glyceraldehydes 3-phosphate (G3P) and dihydroxy-acetone phosphate (DHAP) in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Triosephosphate isomerase
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

TEKNISET TIEDOT

Antigeenin symboli TPI
Antigeenin nimi Triosephosphate isomerase
Environmentally Preferable
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio HRP (Horseradish Peroxidase)
Reaktiivisuus Human,Rat,Mouse
Isäntä Rabbit
Geeni ID 7167
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
Immunogeeni 124-140/140
Lomake liquid
Puhdistus Purified by Protein A
Pitoisuus 1 μg/μl
Ristiadsorptio No
Säilytyspuskuri Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Säilytyslämpötila Store at 4°C for 12 months
Kuljetuslämpötila 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR