Anti-Triosephosphate isomerase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-4042R-A750
BOSSBS-4042R-A750EA 899 EUR
BOSSBS-4042R-A750
Anti-Triosephosphate isomerase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
This gene encodes an enzyme, consisting of two identical proteins, which catalyses the isomerization of glyceraldehydes 3-phosphate (G3P) and dihydroxy-acetone phosphate (DHAP) in glycolysis and gluconeogenesis. Mutations in this gene are associated with triosephosphate isomerase deficiency. Pseudogenes have been identified on chromosomes 1, 4, 6 and 7. Alternative splicing results in multiple transcript variants.

Type: Primary
Antigen: Triosephosphate isomerase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-249/249
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR