Anti-GYG1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-3990R-A680
BOSSBS-3990R-A680EA 890 EUR
BOSSBS-3990R-A680
Anti-GYG1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
This gene encodes a member of the glycogenin family. Glycogenin is a glycosyltransferase that catalyses the formation of a short glucose polymer from uridine diphosphate glucose in an autoglucosylation reaction. This reaction is followed by elongation and branching of the polymer, catalysed by glycogen synthase and branching enzyme, to form glycogen. This gene is expressed in muscle and other tissues. Mutations in this gene result in glycogen storage disease XV. This gene has pseudogenes on chromosomes 1, 8 and 13 respectively. Alternatively spliced transcript variants encoding differentMore Product Information