Anti-NDUFS4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Toimittaja: Bioss
BS-3961R-A350
BOSSBS-3961R-A350EA 893 EUR
BOSSBS-3961R-A350
Anti-NDUFS4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Vasta-aineet
This gene encodes an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), or NADH:ubiquinone oxidoreductase, the first multi-subunit enzyme complex of the mitochondrial respiratory chain. Complex I plays a vital role in cellular ATP production, the primary source of energy for many crucial processes in living cells. It removes electrons from NADH and passes them by a series of different protein-coupled redox centers to the electron acceptor ubiquinone. In well-coupled mitochondria, the electron flux leads to ATP generation via the building of aMore Product Information