Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: Bioss
BS-3756R-HRP
BOSSBS-3756R-HRPEA 900 EUR
BOSSBS-3756R-HRP
Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
PPP1A is a serine/threonine protein phosphatase catalytic subunit that is essential for regulating cellular stress responses in eukaryotes. It binds to magnesium or manganese ions and exists as a monomer. It is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1A is involved in the regulation long term synaptic plasticity and may play an important role in dephosphorylating substrates such as Ca2+/calmodulin dependent protein kinase II.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500