Anti-PPP1CA Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-3756R-A750
BOSSBS-3756R-A750EA 890 EUR
BOSSBS-3756R-A750
Anti-PPP1CA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
PPP1A is a serine/threonine protein phosphatase catalytic subunit that is essential for regulating cellular stress responses in eukaryotes. It binds to magnesium or manganese ions and exists as a monomer. It is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1A is involved in the regulation long term synaptic plasticity and may play an important role in dephosphorylating substrates such as Ca2+/calmodulin dependent protein kinase II.

Type: Primary
Antigen: PPP1CA
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR