Anti-ARA24 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-3751R-A680
BOSSBS-3751R-A680EA 890 EUR
BOSSBS-3751R-A680
Anti-ARA24 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1). Mutations in Ran disrupt DNA synthesis. Because of its many functions, it is likely that Ran interacts with several other proteins. Ran regulates formation and organisation of the microtubule network independently ofMore Product Information