Anti-p70 S6 Kinase Beta Thr444 + Ser447 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-3700R-A750
BOSSBS-3700R-A750EA 917 EUR
BOSSBS-3700R-A750
Anti-p70 S6 Kinase Beta Thr444 + Ser447 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17.
Type: Primary
Antigen: p70More Product Information