Anti-RPS6KB2 Rabbit Polyclonal Antibody
Toimittaja: Bioss
BS-3617R
BOSSBS-3617REA 725 EUR
BOSSBS-3617R
Anti-RPS6KB2 Rabbit Polyclonal Antibody
Vasta-aineet
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based onMore Product Information