Anti-RB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Toimittaja: Bioss
BS-3380R-A555
BOSSBS-3380R-A555EA 898 EUR
BOSSBS-3380R-A555
Anti-RB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Vasta-aineet
Nuclear MarkerRb is a tumor suppressor gene which functions as a negative regulator of the cell cycle by interacting with transcription factors including E2F1, PU1, ATF2, UBF, Elf1 and cAbl. This ability of Rb to alter transcription is regulated by phosphorylation catalyzed by the cyclin dependent protein kinases (cdks). Rb is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus 16E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth.More Product Information