Anti-C3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Toimittaja: Bioss
BS-2934R-A488
BOSSBS-2934R-A488EA 901 EUR
BOSSBS-2934R-A488
Anti-C3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
C3 plays a central role in the activation of the complement system. Its processing by C3 convertase is the central reaction in both classical and alternative complement pathways. After activation C3b can bind covalently, via its reactive thioester, to cell surface carbohydrates or immune aggregates. Derived from proteolytic degradation of complement C3, C3a anaphylatoxin is a mediator of local inflammatory process. In chronic inflammation, acts as a chemoattractant for neutrophils (By similarity). It induces the contraction of smooth muscle, increases vascular permeability and causesMore Product Information