Anti-ACACA Rabbit Polyclonal Antibody (Cy5®)
Toimittaja: Bioss
BS-2745R-CY5
BOSSBS-2745R-CY5EA 900 EUR
BOSSBS-2745R-CY5
Anti-ACACA Rabbit Polyclonal Antibody (Cy5®)
Vasta-aineet
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA.More Product Information