Anti-LIR6 Rabbit Polyclonal Antibody (Cy7®)

Toimittaja: Bioss
BS-2656R-CY7
BOSSBS-2656R-CY7EA 900 EUR
BOSSBS-2656R-CY7
Anti-LIR6 Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
May act as receptor for class I MHC antigens.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIR6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR