Anti-PERK Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Toimittaja: Bioss
BS-2469R-A750
BOSSBS-2469R-A750EA 899 EUR
BOSSBS-2469R-A750
Anti-PERK Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Phosphorylates the alpha subunit of eukaryotic translation-initiation factor 2 (EIF2), leading to its inactivation and thus to a rapid reduction of translational initiation and repression of global protein synthesis. Serves as a critical effector of unfolded protein response (UPR)-induced G1 growth arrest due to the loss of cyclin-D1 (CCND1). Involved in control of mitochondrial morphology and function (By similarity).

Type: Primary
Antigen: PERK
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1001-1116/1116
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

TEKNISET TIEDOT

Antigeenin nimi PERK
Environmentally Preferable
Antigeenin synonyymit PRKR-like endoplasmic reticulum kinase|HsPEK|Pancreatic eIF2-alpha kinase|WRS|PEK|PERK|Eukaryotic translation initiation factor 2-alpha kinase 3|EIF2AK3
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Alexa Fluor® 750
Reaktiivisuus Human,Rat,Mouse
Isäntä Rabbit
Geeni ID 9451
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Virtaussytometria Yes
Modification Unmodified
Epitooppi 1001-1116/1116
Immunogeeni KLH conjugated synthetic peptide derived from human PER
Lomake Liquid
Molekyylipaino 122 kDa
Puhdistus Purified by Protein A
Pitoisuus 1 µg/µl
Säilytyspuskuri Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Säilytyslämpötila Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Kuljetuslämpötila 4 °C
Testatut aplikaatiot ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR