Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Cy5®)

Toimittaja: Bioss
BS-2434R-CY5
BOSSBS-2434R-CY5EA 883 EUR
BOSSBS-2434R-CY5
Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Cy5®)
Vasta-aineet
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of calcium.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP2c1/2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR