Anti-UBF1/RNA polymerase I Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Toimittaja: Bioss
BS-2362R-A680
BOSSBS-2362R-A680EA 890 EUR
BOSSBS-2362R-A680
Anti-UBF1/RNA polymerase I Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Recognises the ribosomal RNA gene promoter and activates transcription mediated by RNA polymerase I through cooperative interactions with the transcription factor SL1/TIF-IB complex. It binds specifically to the upstream control element.

Type: Primary
Antigen: UBF1/RNA polymerase I
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/764
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR